Coomassie® Brillantblau R 250

Lieferant: Cayman Chemical

Synonyme: Brillantblau R, Hydrogen[4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylen]cyclohexa-2,5-dien-1-yliden](ethyl)(m-sulfonatobenzyl)ammonium Mononatriumsalz, Coomassie Blau R 250, Xylenbrillantcyanin, Serva Blue R, Brillantindocyanin 6B, Acid Blue 83

14500-10 14500-25 14500-50 14500-100
CAYM14500-10EA 43 EUR
CAYM14500-10 CAYM14500-25 CAYM14500-50 CAYM14500-100
Coomassie® Brillantblau R 250
Coomassie® Brillantblau R 250
Formel: C₄₅H₄₄N₃NaO₇S₂
Molecular Weight: 825.98 g/mol
Schmelzpunkt: 174…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041762
CAS-Nummer: 6104-59-2
EINECS: 228-060-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR