Virginiamycin complex ≥95%

Lieferant: Cayman Chemical

14503-1 14503-5 14503-10
CAYM14503-1EA 119 EUR
CAYM14503-1 CAYM14503-5 CAYM14503-10
Virginiamycin complex ≥95%
Virginiamycin complex

Virginiamycin complex is a complex containing two streptogramin antibiotics, virginiamycin M1 (75%) and virginiamycin S1 (25%), produced by S. virginiae.


As a complex, the two antibiotics act synergistically to irreversibly inhibit protein synthesis in bacteria. When given to livestock, virginiamycin improves growth. Virginiamycin complex is also used in fuel ethanol fermenters to prevent to control bacteria competing with yeast for nutrients.

Lagertemperatur: Gefrierschrank
CAS-Nummer: 121006-76-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR