D(-)-Luciferin Natriumsalz Monohydrate ≥95%

Lieferant: Cayman Chemical

14682-50 14682-100 14682-500 14682-1
CAYM14682-50EA 50 EUR
CAYM14682-50 CAYM14682-100 CAYM14682-500 CAYM14682-1
D(-)-Luciferin Natriumsalz Monohydrate ≥95%
D(-)-Luciferin Natriumsalz
Formel: C₁₁H₇N₂NaO₃S₂
MDL: MFCD00044938
CAS-Nummer: 103404-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR