Doxorubicin Hydrochlorid ≥98%

Lieferant: Cayman Chemical

Synonyme: Doxorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-10-(((2R,4S,5S,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

15007-5 15007-10 15007-25 15007-50
CAYM15007-5EA 43 EUR
CAYM15007-5 CAYM15007-10 CAYM15007-25 CAYM15007-50
Doxorubicin Hydrochlorid ≥98%
Doxorubicin hydrochloride

Doxorubicin is an anthracycline anti-tumour antibiotic which involves in inhibition of DNA topoisomerase II by inducing double-stranded DNA breaks. It is also involved in the inhibition of nucleic acid synthesis by intercalating within DNA. The accumulation of the p53 tumour suppressor protein leads to the apoptosis in cells.


  • Crystalline solid

Formel: C₂₇H₂₉NO₁₁·HCl
Lagertemperatur: Gefrierschrank
MDL: MFCD00077757
CAS-Nummer: 25316-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR