1,2-Distearoyl-sn-glycero-3-phosphocholine ≥98%
Lieferant: Cayman Chemical
Synonyme:
DSPC, 1,2-Distearoyl-sn-glycero-3-phosphorylcholine, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate
15100-25
CAYM15100-25EA
809
EUR
CAYM15100-25
1,2-Distearoyl-sn-glycero-3-phosphocholine ≥98%
1,2-Distearoyl-sn-glycero-3-phosphocholine
Formel:
C₄₄H₈₈NO₈P Molecular Weight: 790,16 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036905 CAS-Nummer: 816-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...