Lamotrigine ≥98%

Lieferant: Cayman Chemical

Synonyme: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

15428-10 15428-25 15428-100 15428-50
CAYM15428-10EA 76 EUR
CAYM15428-10 CAYM15428-25 CAYM15428-100 CAYM15428-50
Lamotrigine ≥98%
Lamotrigine

Lamotrigine is an inhibitor of voltage-gated sodium channels (Nav), blocking human Nav1,2, Nav1,5, and Nav1,8 with IC50 values of 10, 62, and 96 μM, respectively.


Presumably through this action, lamotrigine acts as an anticonvulsant and has applications in many diseases that feature seizures, including epilepsy and autism spectrum disorder.

Formel: C₉H₇Cl₂N₅
Molecular Weight: 256.09 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00865333
CAS-Nummer: 84057-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR