Imipenem monohydrate
Lieferant: Cayman Chemical
Imipenem is an intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0,5 - 2 µg/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species. Imipenem acts by interfering with bacterial cell wall synthesis.
- A crystalline solid
Formel:
C₁₂H₁₇N₃O₄S Lagertemperatur: Gefrierschrank |
MDL:
MFCD09753321 CAS-Nummer: 74431-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...