Luminol (3-Aminophthalhydrazid) 98

Lieferant: Cayman Chemical
Achtung

Synonyme: NSC 5064

16803-10 16803-25 16803-5 16803-50
CAYM16803-10EA 104 EUR
CAYM16803-10 CAYM16803-25 CAYM16803-5 CAYM16803-50
Luminol (3-Aminophthalhydrazid) 98
Luminol (3-Aminophthalhydrazid)

Luminol is a cyclic diacyl hydrazide that exhibits chemiluminescence upon oxidation.


  • Emission 411 nm
  • Excitation 355 nm
  • Formulation A crystalline solid
  • λmax 220, 295, 355 nm


HRP combined with hydrogen peroxide as an activator causes luminescent peroxidation of luminol. This reaction can be enhanced by certain phenol derivatives, such as p-substituted phenols. Luminol can also be oxidised and chemiluminesce by compounds containing iron, copper, gold and cyanide.

Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00006890
CAS-Nummer: 521-31-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR