Tenofovirdisoproxilfumarat ≥98%, kristalliner Feststoff
Lieferant: Cayman Chemical
|
Gefahr
|
Synonyme:
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate fumarate
Tenofovir disoproxil is a prodrug of the acyclic nucleoside phosphonate, tenofovir. Tenofovir is converted by cellular enzymes to tenofovir diphosphate, an obligate chain terminator that inhibits the activity of HIV reverse transcriptase and hepatitis B virus polymerase. Due to its rapid intracellular uptake, the anti-HIV activity of tenofovir disoproxil is reportedly >100-fold greater than that of the negatively charged tenofovir in a T cell line and primary blood lymphocytes.
- Crystalline solid
- Soluble in organic solvents such as ethanol, DMSO and dimethyl formamide (DMF)
- Sparingly soluble in aqueous buffers
Formel:
C₁₉H₃₀N₅O₁₀P·C₄H₄O₄ Lagertemperatur: Gefrierschrank |
MDL:
MFCD08141829 CAS-Nummer: 202138-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...