Cyclic di-GMP (sodium salt) ≥95%

Lieferant: Cayman Chemical

Synonyme: Cyclic diguanylate, c-di-GMP, 3',5'-Cyclic diguanylic acid, Guanylyl-(3'→5')-3'-guanylic acid, cyclic nucleotide, disodium salt

17144-1 17144-500
CAYM17144-1EA 305 EUR
CAYM17144-1 CAYM17144-500
Cyclic di-GMP (sodium salt) ≥95%
Cyclic di-GMP (sodium salt)

  • A bacterial second messenger and cyclic dinucleotide


A bacterial second messenger and cyclic dinucleotide; an important second messenger in bacteria involved in biofilm formation, motility, virulence, and cell cycling; detected by and binds to STING in eukaryotic cells, activating the innate immune system (Kd = 1,21 µm); induces IFN-β mRNA expression in vitro (EC₅₀= 537,8 nm) less potently than other cyclic dinucleotides.

Formel: C₂₀H₂₂N₁₀O₁₄P₂ ·2Na
Lagertemperatur: Gefrierschrank
CAS-Nummer: 2222132-40-1

Bestellen

Spezifikation Testergebnisse

Formulation A solid
Purity ≥95%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR