Cyclic di-GMP (sodium salt) ≥95%
Lieferant: Cayman Chemical
Synonyme:
Cyclic diguanylate, c-di-GMP, 3',5'-Cyclic diguanylic acid, Guanylyl-(3'→5')-3'-guanylic acid, cyclic nucleotide, disodium salt
17144-1
17144-500
CAYM17144-1EA
305
EUR
CAYM17144-1
CAYM17144-500
Cyclic di-GMP (sodium salt) ≥95%
Cyclic di-GMP (sodium salt)
- A bacterial second messenger and cyclic dinucleotide
A bacterial second messenger and cyclic dinucleotide; an important second messenger in bacteria involved in biofilm formation, motility, virulence, and cell cycling; detected by and binds to STING in eukaryotic cells, activating the innate immune system (Kd = 1,21 µm); induces IFN-β mRNA expression in vitro (EC₅₀= 537,8 nm) less potently than other cyclic dinucleotides.
Formel:
C₂₀H₂₂N₁₀O₁₄P₂ ·2Na Lagertemperatur: Gefrierschrank |
CAS-Nummer:
2222132-40-1 |
Spezifikation Testergebnisse
Formulation | A solid |
Purity | ≥95% |
Stability | ≥ 2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...