trans-2-Decenoic acid ≥98%

Lieferant: Cayman Chemical

17888-10 17888-25 17888-5
CAYM17888-10EA 154 EUR
CAYM17888-10 CAYM17888-25 CAYM17888-5
trans-2-Decenoic acid ≥98%
trans-2-Decenoic acid

trans-2-Decenoic acid is an unsaturated fatty acid found in royal jelly produced from the hypopharyngeal and mandibular gland secretions of honeybees. It is isolated in conjunction with 10-HDA and both demonstrate weak estrogenic activity, inhibiting binding of 17β-estradiol to estrogen receptor β.

Formel: C₁₀H₁₈O₂
MDL: MFCD00039530
CAS-Nummer: 334-49-6

Bestellen

Spezifikation Testergebnisse

Purity ≥98%
Formulation A neat oil
λmax 209 nm
Storage –20 °C
Stability ≥1 year


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR