BI-605906 ≥98%

Lieferant: Cayman Chemical

19184-1 19184-5
CAYM19184-1EA 101 EUR
CAYM19184-1 CAYM19184-5
BI-605906 ≥98%
BI-605906

  • An IKKβ inhibitor


An IKKβ inhibitor (IC50 = 380 nM); selective for IKKβ over insulin-like growth factor 1 (IGF1; IC50 = 7.6 μM) and over 100 kinases in a panel up to 10 μM; partially inhibits IL-1-stimulated activation of IKKε/TBK1 in vitro; inhibits TNF-α-dependent IkB degradation and expression of the proinflammatory cytokines IL-6, IL-1β, and CXCL1/2 without affecting lipogenic gene expression or glucose production in murine primary hepatocytes

Formel: C₁₇H₂₂F₂N₄O₃S₂
Lagertemperatur: Gefrierschrank
CAS-Nummer: 960293-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a199494492ff9b45fa6f0969d0c8750c0a656e9&vid=ca11e83e690b47d0f92510810c22913d677afac0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR