Sisomicin ≥98%

Lieferant: Cayman Chemical
Gefahr

Synonyme: Rickamicin, Salvamina, SCH 13475, Antibiotic 6640

21046-1 21046-100 21046-50 21046-500
CAYM21046-1EA 405 EUR
CAYM21046-1 CAYM21046-100 CAYM21046-50 CAYM21046-500
Sisomicin ≥98%
Sisomicin

Sisomicin is a broad-spectrum aminoglycoside antibiotic originally isolated from M. inyoensis.


It is active against Gram-positive and Gram-negative bacteria including B. subtilis, S. aureus, E. coli, and P. aeruginosa among others (MICs = 0.01-0.75 µg/ml) but is less active against P. multocida and V. coli (MICs = 7.5 and 2.4 µg/ml, respectively). Sisomicin is protective against Gram-positive and Gram-negative bacterial infections in mice with 50% protective dose (PD50s) values ranging from 0.5 to 6 mg/kg.

Formel: C₁₉H₃₇N₅O₇
Molecular Weight: 447.50 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00865123
CAS-Nummer: 32385-11-8
EINECS: 251-018-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR