Human recombinant STING A162 variant (from E. coli)
Lieferant: Cayman Chemical
STING A162 variant (human recombinant) contains amino acids 138-379 of the wild-type variant (R232) with an alanine substituted for serine at position 162. Stimulator of interferon genes (STING) is a component of the innate immune response that binds to cyclic dinucleotides, which are bacterial second messengers, leading to activation of NF-κB and transcription of immunomodulatory genes, including type I interferon (IFN). The R232 variant of STING is the most common variant in the human population, found at a frequency of 57,9% in the 1000 Genome Project. The SNP variant H232 is found at a 13,7% frequency. The S162A point mutation is located in the cyclic dinucleotide binding site of the human STING variants R232 and H232 and allows human STING to bind to DMXAA, a compound previously known to bind mouse, but not human, STING. When STING S162A is bound to DMXAA, it adopts the closed conformation, similar to the conformation it has when bound to the second messenger 2’3’-cGAMP (Item No. 19887), and activates the IFN pathway similarly to mouse STING.
Used for: Antivirals, Autoimmunity, Innate Immunity