BSA-Palmitate Saturated Fatty Acid Complex
Lieferant: Cayman Chemical
BSA-Palmitate saturated fatty acid complex is composed of palmitic acid and bovine serum albumin (BSA) at an approximately 6:1 molar ratio of palmitate:BSA. It can be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring fatty acid oxidation or similar processes in various cellular metabolic studies.
- A fatty acid substrate
- Origin: Animal/Bovine
- Stability: ≥ 1 year
The BSA/BSA-FAs are acceptable for use with short-term cell culture applications (acute treatment to 18 hours), however for long-term applications (25+ hours) the product can/should be filter sterilized using a 0.2 µm filter and sterile receptacle without affecting the performance. For best results, it is recommended that this product be used in conjunction with BSA control for fatty acid complexes (5 mM).
Formulation: 5 mM Palmitate:0.8 mM BSA (6:1 palmitate:BSA) in 150 mM sodium chloride, pH 7.4
Bitte beachten Store at –20 °C.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...