Apiin ≥98%

Lieferant: Cayman Chemical

Synonyme: Apigenin-7-Apioglucoside, 7-(((2S,3R,4S,5S,6R)-3-(((2S,3R,4R)-3,4-Dihydroxy-4-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

34596-50
CAYM34596-50EA 79800 EUR
CAYM34596-50
Apiin ≥98%
Apiin
Formel: C₆H₁₀N₄O₂
Molecular Weight: 564,49 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00016941
CAS-Nummer: 26544-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR