N-(3-Benzylthiazol-2(3H)-ylidene)-1H-pyrrolo[2,3-b]pyridine-3-carboxamide ≥98%

Lieferant: Cayman Chemical
Achtung

Synonyme: TRULI

36623-1
CAYM36623-1EA 39 EUR
CAYM36623-1
N-(3-Benzylthiazol-2(3H)-ylidene)-1H-pyrrolo[2,3-b]pyridine-3-carboxamide ≥98%
N-(3-Benzylthiazol-2(3H)-ylidene)-1H-pyrrolo[2,3-b]pyridine-3-carboxamide
Formel: C₁₈H₁₄N₄OS
Molecular Weight: 334,3950 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD33402276
CAS-Nummer: 1424635-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR