9(R)-HODE 0,01% in Ethanol , 98%

Lieferant: Cayman Chemical

38405-25 38405-50 38405-100 38405-250
CAYM38405-25EA 108 EUR
CAYM38405-25 CAYM38405-50 CAYM38405-100 CAYM38405-250
9(R)-HODE 0,01% in Ethanol , 98%
9(R)-HODE

9(R)-HODE is one of several monohydroxylated products of linoleic acid. All known mammalian lipoxygenases appear to catalyze the oxygenation of arachidonic and linoleic acid to give products having strictly the (S) configuration at the site of oxygen insertion. However, both human umbilical vein endothelial cells and bovine aorta endothelial cells have been shown to produce 9(R)-HODE when incubated with linoleic acid.
The physiological function of 9(R)-HODE and the enzyme that catalyzes its formation have not been determined.


  • Purity: ≥98%
  • Formulation: A solution in ethanol
  • λmax: 234 nm


Warning - this product is not for human or veterinary use.

Formel: C₁₈H₃₂O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00084832
CAS-Nummer: 10075-11-3
UN: 1170
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR