9(S)-HODE ≥98%

Lieferant: Cayman Chemical

38410-1 38410-100 38410-500
CAYM38410-1EA 1223 EUR
CAYM38410-1 CAYM38410-100 CAYM38410-500
9(S)-HODE ≥98%
9(S)-HODE

9(S)-HODE is produced by the lipoxygenation of linoleic acid in both plants and animals. It has been detected in atherosclerotic plaques, as an esterified component of membrane phospholipids and in oxidised LDL particles.


9(S)-HODE is supplied as a solution in ethanol. To change the solvent, simply evaporate the ethanol under a gentle stream of nitrogen and immediately add the solvent of choice. Solvents such as DMSO and dimethyl formamide purged with an inert gas can be used. The solubility of 9(S)-HODE in these solvents is approximately 50 mg/ml.


An organic solvent-free solution of 9(S)-HODE can be prepared by evaporating the ethanol and directly dissolving the neat oil in aqueous buffers. The solubility of 9(S)-HODE in PBS (pH 7,2) is approximately 1 mg/ml. Not recommend to store the aqueous solution for more than one day.

Formel: C₁₈H₃₂O₃
MDL: MFCD00065831
CAS-Nummer: 73543-67-6

Bestellen

Spezifikation Testergebnisse

Purity ≥98%
Stability ≥1 year at −20 °C
Formulation A Solution in Ethanol
UV/Vis λmax: 234 nm ε: 23000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR