BSA-Linoleate ω-6 Polyunsaturated Fatty Acid Complex

Lieferant: Cayman Chemical

38649-5
CAYM38649-5EA 68 EUR
CAYM38649-5
BSA-Linoleate ω-6 Polyunsaturated Fatty Acid Complex
Standards Lebensmittel-Standards

BSA-linoleate ω-6 polyunsaturated fatty acid complex is composed of linoleic acid and bovine serum albumin (BSA) at an approximately 6:1 molar ratio of linoleate:BSA.


  • Source: Animal/bovine
  • Formulation: 1 mM linoleate:0,17 mM BSA in 150 mM sodium chloride
  • Storage: –20 °C
  • Stability: ≥1 year


BSA-linoleate complexes can be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring fatty acid oxidation or similar processes in various cellular metabolic studies. The BSA/BSA-FAs are acceptable for use with short-term cell culture applications (acute treatment to 18 hours), however, for long-term applications (25+ hours) the product can/should be filter-sterilised using a 0,2 µm filter and sterile receptacle without affecting the performance.


For best results, it is recommended that this product be used in conjunction with BSA control for BSA-fatty acid complexes (1 mM).

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR