Ferroptocide ≥90%

Lieferant: Cayman Chemical

Synonyme: (2S,3aS,4S,5R,7R,8S,8aS,9R)-octahydro-2,8a-dihydroxy-4,8,9-trimethyl-1-oxo-5-(2,3,5,8-tetrahydro-1,3-dioxo-2-phenyl-1H-[1,2,4]triazolo[1,2-a]pyridazin-6-yl)-1H-3a,8-propanoazulen-7-yl ester, 2-chloro-acetic acid

39005-5
CAYM39005-5EA 178 EUR
CAYM39005-5
Ferroptocide ≥90%
Ferroptocide
Formel: C₃₀H₃₆ClN₃O₇
Molecular Weight: 586,08 g/mol
Lagertemperatur: Room Temperature
CAS-Nummer: 2505218-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR