1-(5-Chloro-2-hydroxyphenyl)ethanone (2,4,6-trichlorophenyl)hydrazone ≥95%

Lieferant: Cayman Chemical

Synonyme: Mitochondrial fusion promoter M1, (E)-4-Chloro-2-(1-(2-(2,4,6-trichlorophenyl)hydrazono)ethyl)phenol

39909-10
CAYM39909-10EA 64 EUR
CAYM39909-10
1-(5-Chloro-2-hydroxyphenyl)ethanone (2,4,6-trichlorophenyl)hydrazone ≥95%
1-(5-Chloro-2-hydroxyphenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
Formel: C₁₄H₁₀Cl₄N₂O
Molecular Weight: 364,054 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00116814
CAS-Nummer: 219315-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR