O-Methylhydroxylamin Hydrochlorid
Lieferant: Cayman Chemical
400036-100
400036-500
CAYM400036-100EA
16
EUR
CAYM400036-100
CAYM400036-500
O-Methylhydroxylamin Hydrochlorid
O-Methylhydroxylamin Hydrochlorid
Formel:
CH₆ClNO |
MDL:
MFCD00012951 CAS-Nummer: 593-56-6 EINECS: 209-798-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...