O-Methylhydroxylamin Hydrochlorid

Lieferant: Cayman Chemical

400036-100 400036-500
CAYM400036-100EA 16 EUR
CAYM400036-100 CAYM400036-500
O-Methylhydroxylamin Hydrochlorid
O-Methylhydroxylamin Hydrochlorid
Formel: CH₆ClNO
MDL: MFCD00012951
CAS-Nummer: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR