(±)11(12)-EET methyl ester-Lösung ≥98%
Lieferant: Cayman Chemical
|
Gefahr
|
(±)11(12)-EET is biosynthesized in rat and rabbit liver microsomes by CYP450. (±)11(12)-EET has been shown, along with (±)8(9)-EET, to play a role in the recovery of depleted Ca²⁺ pools in cultured smooth muscle cells.
(±)11(12)-EET methyl ester is a more stable for used for long-term storage. It can be readily hydrolysed to the free acid as needed.
Formel:
C₂₁H₃₄O₃ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077342 CAS-Nummer: 73799-06-1 UN: 1170 ADR: 3,II |
Spezifikation Testergebnisse
Purity | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...