Ölrot O-Lösung Lipidtropfen-Analyse

Lieferant: Cayman Chemical
Gefahr

Synonyme: CI 26125

600045-25
CAYM600045-25EA 34 EUR
CAYM600045-25
Ölrot O-Lösung Lipidtropfen-Analyse
Ölrot O

  • Formulation: A 25 ml stock solution

Formel: C₂₆H₂₄N₄O
Molecular Weight: 408,51 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00003898
CAS-Nummer: 1320-06-5
UN: 1219
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR