Nilrot -Lösung in DMSO
Lieferant: Cayman Chemical
|
Achtung
|
Synonyme:
9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
600055-1
CAYM600055-1EA
161
EUR
CAYM600055-1
Nilrot -Lösung in DMSO
Nilrot
Cell-based working solution prepared by dilution of the Nile red stock solution 1:1000 in the assay buffer.
- Mix to make sure no particles or flakes remain
- Tested and formulated for lipid droplets fluorescence assay kits
Designed for applications in angiogenesis, apoptosis and cancer research.
Bitte beachten Not perform as described if used with other assay reagents or protocols − Not for human or veterinary use.
Formel:
C₂₀H₁₈N₂O₂ |
MDL:
MFCD00011639 CAS-Nummer: 7385-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...