Anti-Ovalbumin IgG1 (Mouse) ELISA Kit
Lieferant: Cayman Chemical
For the measurement of anti-ovalbumin of the IgG1 isotype in mouse plasma and serum.
- Measure mouse anti-OVA IgG1 in mouse plasma, serum, or BAL fluids
- Employs an affinity-purified mouse anti-OVA IgG1 polyclonal antibody standard for accurate results
- Assay Range: 1.56 – 200 ng/ml with an LLOQ of 1.56 ng/ml
- Rapid assay; get results in under 4 hours
- LLOD: 0.006 U/ml
Cayman's Ovalbumin IgG1 (mouse) ELISA Kit is an immunometric (i.e.,'sandwich') assay which can be used to measure anti-ovalbumin of the IgG1 isotype in mouse plasma and serum without prior sample purification.