Hemoglobin standard
Lieferant: Cayman Chemical
700543-1
CAYM700543-1EA 23 EUR
CAYM700543-1
Hemoglobin standard
Standards Biologische Standards
Ready for use with colorimetric assay kit. This vial contains solution of 500 µM heme.
Stability: ≥1 year at 4 °C.
Bitte beachten For laboratory use only. Not for human or veterinary use.