2-Fluor-α-methyl-4-biphenylessigsäure

Lieferant: Cayman Chemical

Synonyme: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

70250-1 70250-5 70250-10 70250-25
CAYM70250-1EA 43 EUR
CAYM70250-1 CAYM70250-5 CAYM70250-10 CAYM70250-25
2-Fluor-α-methyl-4-biphenylessigsäure
2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
Molecular Weight: 244.265 g/mol
MDL: MFCD00079303
CAS-Nummer: 5104-49-4
EINECS: 225-827-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR