2-Fluor-α-methyl-4-biphenylessigsäure
Lieferant: Cayman Chemical
Synonyme:
Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys
70250-1
70250-5
70250-10
70250-25
CAYM70250-1EA
43
EUR
CAYM70250-1
CAYM70250-5
CAYM70250-10
CAYM70250-25
2-Fluor-α-methyl-4-biphenylessigsäure
2-Fluor-α-methyl-4-biphenylessigsäure
Formel:
C₁₅H₁₃FO₂ |
MDL:
MFCD00079303 CAS-Nummer: 5104-49-4 EINECS: 225-827-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...