N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid

Lieferant: Cayman Chemical
Achtung

70455-5 70455-10 70455-25 70455-50
CAYM70455-5EA 46 EUR
CAYM70455-5 CAYM70455-10 CAYM70455-25 CAYM70455-50
N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid
N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid
Formel: C₁₀H₁₈Cl₂N₂
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012482
CAS-Nummer: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e955f6b2a1434ddba239f533c6611583219f8904&vid=13fb110b013db70c026f8eefeee7f08fa2568c0c&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR