Ebselen

Lieferant: Cayman Chemical
Gefahr

Synonyme: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

70530-100 70530-5 70530-50 70530-25
CAYM70530-100EA 308 EUR
CAYM70530-100 CAYM70530-5 CAYM70530-50 CAYM70530-25
Ebselen
Ebselen
Formel: C₁₃H₉NOSe
Molecular Weight: 274.20 g/mol
MDL: MFCD00210937
CAS-Nummer: 60940-34-3
UN: 3283
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR