Resveratrol

Lieferant: Cayman Chemical

70675-100 70675-50 70675-500 70675-250
CAYM70675-100EA 60 EUR
CAYM70675-100 CAYM70675-50 CAYM70675-500 CAYM70675-250
Resveratrol
Resveratrol
Formel: C₁₄H₁₂O₃
Schmelzpunkt: 260 °C
Dichte: 1,36 g/cm³ (20 °C)
Lagertemperatur: Gefrierschrank
MDL: MFCD00133799
CAS-Nummer: 501-36-0
Merck Index: 12,08326

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR