Sterile burn gel dressing
Lieferant: ORKLA CARE
51011015
CEDE51011015EA 117 EUR
CEDE51011015
Sterile burn gel dressing
Erste-Hilfe-Ausstattung Brandwundenversorgung
Burn gel dressing provides rapid cooling and effective pain relief for first and second degree burns.
  • Sterile dressing prevents injury from penetrating deeper into skin
  • Cold reduces pain and swelling
  • Shelf life of 5 years
Easy to use with clear instructions. Suitable for use on burns up to 20x20 cm. For larger areas use several dressings.