Wandbefestigung für Augenspülung
Lieferant: ORKLA CARE
7202
CEDE7202EA 0 EUR
CEDE7202
Wandbefestigung für Augenspülung
Wandhaken
Zwei Halterungen aus grünem Kunststoff für die Lagerung von zwei Flaschen Cederroth-Augenspülung (500 ml).
Holders are fastened on a metal plate, which can be mounted on any vertical surface example on a wall.
Also serves as an opening device for the eyewash bottles: Bottle is automatically opened when removed from the holder.
Afterglowing sign with instructions for use is included.