IBC bunds, PE
Lieferant: CEMO
8582
CEMG8582EA 1560 EUR
CEMG8582
IBC bunds, PE
Absorptionsmaterial
IBC bunds are manufactured from high quality polyethylene (HD-LMD).
  • With general construction inspection approval no.: Z-40.22-451
  • High resistance to chemicals
  • Accessible to forklift or walkietruck