Drum filling station
Lieferant: CEMO
8674
CEMG8674EA 597 EUR
CEMG8674
Drum filling station
Fasslager
This filling station is suitable as a storage and filling station for two 200 L barrels and with additional drum pallet for four 200 L drums.
  • Made from high-quality polyethylene
  • High chemical resistance
  • Can be driven under
  • Collection volume: 450 L