icanCLAVE STE-45-T Autocalve and CertoAssist - Set
Lieferant: CERTOCLAV
8500610
CERT8500610EA 10330 EUR
CERT8500610
icanCLAVE STE-45-T Autocalve and CertoAssist - Set
Sterilisatoren Autoklaven (Dampfsterilisatoren)
The STE-45-T autoclave makes everyday laboratory work much easier thanks to its predefined standard programmes, automatic documentation and short sterilisation cycles. Its preheatable chamber, the air cooling system and the vacuum pump enable a fast sterilisation cycle including drying.
  • 11 preinstalled programs, freely editable, up to 50 user-definable programs in three different types (solid, liquid, vacuum test) can be freely created
  • Flexible media sensor PT1000
  • Water quality sensor
  • 7" touchscreen with video assistant
  • Automatic documentation
  • RS232, RS485,
More Product Information