Natriumdihydrogenphosphat Monohydrat 98,0-103,0% USP, Multikompendial, J.T.Baker®

Lieferant: Avantor

Synonyme: Natriumphosphat monobasisch Monohydrat

Veränderungsmanagement-Kategorie= HR

CH04-07EA 0 EUR
CH04-07
Natriumdihydrogenphosphat Monohydrat 98,0-103,0% USP, Multikompendial, J.T.Baker®
Natriumdihydrogenphosphat Monohydrat
Formel: NaH₂PO₄·H₂O
Molecular Weight: 137,99 g/mol
Schmelzpunkt: 100 °C
Dichte: 2,04 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets ChP Chemical Specifications
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Appearance (White crystals or granules) Passes Test
USP - Assay (anhydrous basis) 98.0 - 103.0 %
USP - Aluminum, Calcium, and Related Elements Passes Test
USP - Arsenic (As) ≤ 8 ppm
USP - Chloride (Cl) ≤ 0.014 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances ≤ 0.2 %
USP - pH (1 in 20) 4.1 - 4.5
USP - Sulfate (SO₄) ≤ 0.15 %
USP - Water (H₂O) 10.0 - 15.0 %
EP - Identification A Passes Test
EP - Identification B Passes Test
EP - Identification C Passes Test
EP - Assay (anhydrous basis) 98.0 - 100.5 %
EP - Chloride (Cl) ≤ 200 ppm
EP - Sulfate (SO₄) ≤ 300 ppm
EP - Reducing Substances Passes Test
Endotoxin Concentration (EU/g) ≤ 2.5
EP - Iron (Fe) ≤ 10 ppm
EP - Appearance of Solution Passes Test
EP - pH 4.2 - 4.5
BP - Identification A Passes Test
BP - Identification B Passes Test
BP - Identification C Passes Test
BP - Identification D Passes Test
BP - Acidity 4.2 - 4.5
BP - Clarity and Color of Solution Passes Test
BP - Arsenic (As) ≤ 2 ppm
BP - Iron (Fe) ≤ 10 ppm
BP - Chloride (Cl) ≤ 200 ppm
BP - Sulfate (SO₄) ≤ 300 ppm
BP - Reducing Substances Passes Test
BP - Loss on Drying at 130°C 11.5 - 14.5 %
BP - Assay (dried basis) 98.0 - 100.5 %
JPE - Assay (dried basis) ≥ 98.0 %
JPE - Identification Passes Test
JPE - pH 4.1 - 4.7
JPE - Clarity and Color of Solution Passes Test
JPE - Chloride (Cl) ≤ 0.005 %
JPE - Sulfate (SO4) ≤ 0.019 %
JPE - Heavy Metals ≤ 10 ppm
JPE - Arsenic (As) ≤ 2 ppm
ChP - Assay (dried basis) ≥ 98.0 %
ChP - Identification Passes Test
ChP - Acidity (pH) 4.1 - 4.7
ChP - Clarity and color of solution Passes Test
ChP - Chlorides ≤ 0.01 %
ChP - Sulfates ≤ 0.05 %
ChP - Loss on Drying 10.0 - 15.0 %
ChP - Aluminum ≤ 0.01 %
ChP - Calcium ≤ 0.01 %
ChP - Heavy metals ≤ 0.001 %
ChP - Arsenic ≤ 0.0002 %
ChP - Water insoluble substanc ≤ 0.2 %
ChP - Reducing substances Passes Test
ChP - Iron ≤ 0.001 %
For EP notated requirements, results are derived from testing Sodium
Phosphate, Monobasic Monohydrate, U.S.P. to the EP Chemical
specifications for Sodium Dihydrogen Phosphate Dihydrate.
For JPE notated requirements, results are derived from testing Sodium
Phosphate, Monobasic Monohydrate, U.S.P. to the JPE Chemical
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR