Glycin (H-Gly-OH)
Lieferant: CHATTEM CHEMICALS
Synonyme:
H-Gly-OH, Glycocoll, Aminoessigsäure
BD2128U.9012
CHATBD2128U.9012EA
0
EUR
CHATBD2128U.9012
Glycin (H-Gly-OH)
Glycin (H-Gly-OH)
Formel:
NH₂CH₂COOH Molecular Weight: 75.07 g/mol Siedepunkt: 181 Schmelzpunkt: 83 Dichte: 1.64 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008131 CAS-Nummer: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...