Visual colour kits, dissolved oxygen, CHEMets®
Lieferant: CHEMetrics
K-7501
CHETK-7501EA 355 EUR
CHETK-7501
Visual colour kits, dissolved oxygen, CHEMets®
Umwelt-Prüfkits
Ideal for low- to medium-range colorimetric analysis of water quality
CHEMets® test kits utilise self-filling ampules and vacuum pulls the sample up from a snap cup into the ampule when the tip is broken off. After tilting the ampule several times to mix the sample and reagent and waiting for the colour to develop (usually 2 minutes or less), the analyst quantifies the result using the provided cylindrical comparator.
Lieferumfang: Kit comes in a plastic case and contains everything needed to perform 30 tests: 30 ampoules, comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube, 25 ml sample cup and instructions.