Natrium-1-decansulfonat

Lieferant: CHEMPUR

Synonyme: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

002016.25G
CHMP002016.25GEA 0 EUR
CHMP002016.25G
Natrium-1-decansulfonat
Natrium-1-decansulfonat
Formel: C₁₀H₂₁NaO₃S
Molecular Weight: 244,33 g/mol
Schmelzpunkt: > 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007526
CAS-Nummer: 13419-61-9
EINECS: 236-525-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR