Erbiumtrinitrat Pentahydrat

Lieferant: CHEMPUR

Synonyme: Erbium(III)nitrat Pentahydrat

003018.0025
CHMP003018.0025EA 0 EUR
CHMP003018.0025
Erbiumtrinitrat Pentahydrat
Erbiumtrinitrat Pentahydrat
Formel: ErN₃O₉·5H₂O
MDL: MFCD00149691
CAS-Nummer: 10031-51-3
EINECS: 233-436-7
UN: 1477
ADR: 5.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR