Digalliumtrioxid

Lieferant: CHEMPUR

003827.050
CHMP003827.050EA 0 EUR
CHMP003827.050
Digalliumtrioxid
Digalliumtrioxid
Formel: Ga₂O₃
Molecular Weight: 187.44 g/mol
Schmelzpunkt: 1794.85
Dichte: 5.95 g/cm³ (27 °C)
MDL: MFCD00011020
CAS-Nummer: 12024-21-4
EINECS: 234-691-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR