Indium(III)oxid
Lieferant: CHEMPUR
Synonyme:
Diindiumtrioxid, Indiumtrioxid
004424.025
CHMP004424.025EA
0
EUR
CHMP004424.025
Indium(III)oxid
Indium(III)oxid
Formel:
In₂O₃ Molecular Weight: 277,64 g/mol Schmelzpunkt: 715,05 °C Dichte: 7,18 g/cm³ (20 °C) |
MDL:
MFCD00011060 CAS-Nummer: 1312-43-2 EINECS: 215-193-9 Merck Index: 12,04984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...