Terbiumtrinitrat Hydrat

Lieferant: CHEMPUR

007233.0010
CHMP007233.0010EA 0 EUR
CHMP007233.0010
Terbiumtrinitrat Hydrat
Terbiumtrinitrat Hydrat
Formel: N₃O₉Tb·xH₂O
Schmelzpunkt: 89,3 °C
MDL: MFCD00011257
CAS-Nummer: 10043-27-3
EINECS: 233-138-7
UN: 1477
ADR: 5.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR