Ytterbium(III) nitrate hydrate

Lieferant: CHEMPUR

Synonyme: Ytterbium trinitrate hydrate

008309.0025
CHMP008309.0025EA 0 EUR
CHMP008309.0025
Ytterbium(III) nitrate hydrate
Ytterbium(III) nitrate hydrate
Formel: Yb(NO₃)₃·xH₂O
Molecular Weight: 359.05 g/mol (anhy)
MDL: MFCD00149883
EINECS: 237-384-6
UN: 1477
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR