Platin 99,99%, Drahtform Ø 0,5 mm

Lieferant: CHEMPUR

Synonyme: Platinium

009345-1M
CHMP009345-1MEA 0 EUR
CHMP009345-1M
Platin 99,99%, Drahtform Ø 0,5 mm
Platinum
Formel: Pt
Molecular Weight: 195.08 g/mol
Siedepunkt: 3827
Schmelzpunkt: 1769
MDL: MFCD00011179
CAS-Nummer: 7440-06-4
Merck Index: 12,07689

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR